Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPS3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100370.3 | NP_171984.2 | 9 - 10 | 2976 | 86 | |
GenBank mRNA | AY078949.1 | - | 9 - 10 | 2887 | 86 | |
BT002210.1 | - | 9 - 10 | 2779 | 86 | ||
KJ139031.1 | - | 9 - 10 | 2779 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |