Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP88A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331521.1 | NP_001320198.1 | 2 - 3 | 574 | 75 | |
NM_100394.4 | NP_172008.1 | 2 - 3 | 641 | 75 | ||
GenBank mRNA | AK229009.1 | - | 2 - 3 | 576 | 75 | |
BT029169.1 | - | 1 - 2 | 240 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |