Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197984.1 | NP_001184913.1 | 35 - 36 | 3978 | 82 | |
NM_001331555.1 | NP_001322482.1 | 34 - 35 | 4192 | 82 | ||
NM_001331556.1 | NP_001322483.1 | 33 - 34 | 4026 | 82 | ||
NM_100436.5 | NP_563743.2 | 34 - 35 | 4275 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |