Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331626.1 | NP_001321476.1 | 2 - 3 | 2254 | 60 | |
NM_001331627.1 | NP_001321475.1 | 2 - 3 | 2333 | 60 | ||
NM_100521.3 | NP_172129.1 | 2 - 3 | 2256 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |