Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100531.3 | NP_563768.1 | 1 - 2 | 1171 | 84 | |
GenBank mRNA | AK229141.1 | - | 1 - 2 | 926 | 84 | |
BT033096.1 | - | 1 - 2 | 883 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |