Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100592.5 | NP_563783.1 | 4 - 5 | 1228 | 63 | |
GenBank mRNA | AK118982.1 | - | 4 - 5 | 1012 | 63 | |
BT005564.1 | - | 4 - 5 | 918 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |