Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160845.2 | NP_001154317.1 | 2 - 3 | 414 | 122 | |
NM_001160846.1 | NP_001154318.1 | 2 - 3 | 462 | 122 | ||
NM_001331697.1 | NP_001322741.1 | 2 - 3 | 733 | 122 | ||
NM_001331698.1 | NP_001322740.1 | 2 - 3 | 733 | 122 | ||
NM_100613.5 | NP_172219.5 | 2 - 3 | 462 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |