Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112950.3 | NP_188694.2 | 2 - 3 | 1311 | 96 | |
GenBank mRNA | AK118605.1 | - | 2 - 3 | 1226 | 96 | |
BT005952.1 | - | 2 - 3 | 1079 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |