Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPFTSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130140.3 | NP_566056.1 | 3 - 4 | 647 | 69 | |
GenBank mRNA | AF360125.1 | - | 3 - 4 | 517 | 69 | |
AJ010820.1 | - | 3 - 4 | 497 | 69 | ||
AY051026.1 | - | 3 - 4 | 487 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |