Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084596.2 | NP_001078065.1 | 8 - 9 | 1450 | 104 | |
NM_001337163.1 | NP_001325355.1 | 8 - 9 | 1662 | 104 | ||
NM_001337164.1 | NP_001325353.1 | 7 - 8 | 1401 | 104 | ||
NM_001337165.1 | NP_001325354.1 | 7 - 8 | 1401 | 104 | ||
NM_130158.4 | NP_566058.2 | 8 - 9 | 1598 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |