Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130253.3 | NP_182209.1 | 4 - 5 | 403 | 68 | |
GenBank mRNA | BT025278.1 | - | 3 - 4 | 343 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |