Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MHX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130328.3 | NP_566105.1 | 4 - 5 | 1023 | 65 | |
GenBank mRNA | AF109178.1 | - | 4 - 5 | 856 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |