Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125072.1 | NP_001118544.1 | 10 - 11 | 940 | 149 | |
NM_130361.5 | NP_566115.1 | 10 - 11 | 993 | 149 | ||
GenBank mRNA | AF245171.1 | - | 10 - 11 | 924 | 149 | |
AF326865.1 | - | 10 - 11 | 866 | 149 | ||
AF349516.1 | - | 10 - 11 | 856 | 149 | ||
AY075700.1 | - | 10 - 11 | 866 | 149 | ||
AY102139.1 | - | 10 - 11 | 856 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |