Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMPK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035913.2 | NP_001030990.1 | 5 - 6 | 1077 | 92 | |
NM_100662.4 | NP_172266.2 | 4 - 5 | 1182 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |