Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331741.1 | NP_001318950.1 | 22 - 23 | 2867 | 100 | |
NM_100665.3 | NP_172269.2 | 22 - 23 | 2745 | 100 | ||
GenBank mRNA | AK230326.1 | - | 22 - 23 | 2712 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |