Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100711.4 | NP_563815.1 | 2 - 3 | 345 | 67 | |
GenBank mRNA | AJ493060.1 | - | 2 - 3 | 181 | 67 | |
AY050960.1 | - | 2 - 3 | 174 | 67 | ||
AY087368.1 | - | 2 - 3 | 239 | 67 | ||
AY091345.1 | - | 2 - 3 | 150 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |