Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100727.4 | NP_172330.1 | 6 - 7 | 1597 | 77 | |
GenBank mRNA | AB004293.1 | - | 6 - 7 | 1553 | 77 | |
AB004820.1 | - | 6 - 7 | 1518 | 77 | ||
BT030346.1 | - | 6 - 7 | 1431 | 77 | ||
Y15362.1 | - | 1482 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |