Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035923.3 | NP_001031000.1 | 4 - 5 | 708 | 92 | |
NM_001331777.1 | NP_001321301.1 | 3 - 4 | 1071 | 92 | ||
NM_100728.4 | NP_172331.1 | 4 - 5 | 776 | 92 | ||
GenBank mRNA | AF370251.1 | - | 4 - 5 | 632 | 92 | |
AK317253.1 | - | 4 - 5 | 551 | 92 | ||
AY063067.1 | - | 3 - 4 | 477 | 92 | ||
U44133.1 | - | 3 - 4 | 521 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |