Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035929.1 | NP_001031006.1 | 4 - 5 | 573 | 62 | |
NM_100765.5 | NP_563830.1 | 4 - 5 | 738 | 62 | ||
GenBank mRNA | AJ249967.1 | - | 4 - 5 | 530 | 62 | |
BT008661.1 | - | 4 - 5 | 548 | 62 | ||
D89051.1 | - | 4 - 5 | 473 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |