Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100773.4 | NP_563834.1 | 3 - 4 | 1039 | 75 | |
GenBank mRNA | AF439826.1 | - | 3 - 4 | 992 | 75 | |
BT000625.1 | - | 3 - 4 | 389 | 75 | ||
DQ132632.1 | - | 3 - 4 | 992 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |