Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACLA-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100814.4 | NP_172414.1 | 4 - 5 | 663 | 87 | |
GenBank mRNA | AY127019.1 | - | 4 - 5 | 555 | 87 | |
BT003017.1 | - | 4 - 5 | 524 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |