Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035938.2 | NP_001031015.1 | 7 - 8 | 810 | 57 | |
NM_001035939.3 | NP_001031016.2 | 6 - 7 | 920 | 57 | ||
NM_100881.3 | NP_172478.1 | 6 - 7 | 894 | 57 | ||
GenBank mRNA | AF370135.1 | - | 6 - 7 | 891 | 57 | |
AJ271731.1 | - | 6 - 7 | 778 | 57 | ||
AK317528.1 | - | 7 - 8 | 811 | 57 | ||
AK319062.1 | - | 6 - 7 | 917 | 57 | ||
AY051038.1 | - | 6 - 7 | 726 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |