Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100887.3 | NP_563860.1 | 21 - 22 | 2067 | 91 | |
GenBank mRNA | AK229199.1 | - | 21 - 22 | 2067 | 91 | |
EU082212.1 | - | 21 - 22 | 1878 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |