Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331906.1 | NP_001321764.1 | 17 - 18 | 3471 | 127 | |
NM_001331907.1 | NP_001321765.1 | 16 - 17 | 3258 | 127 | ||
NM_100919.5 | NP_172515.2 | 17 - 18 | 3471 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |