Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198028.1 | NP_001184957.1 | 1 - 2 | 351 | 79 | |
NM_100981.4 | NP_172575.2 | 1 - 2 | 556 | 79 | ||
GenBank mRNA | BT030381.1 | - | 1 - 2 | 302 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |