Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100998.4 | NP_172592.1 | 2 - 3 | 682 | 62 | |
GenBank mRNA | AY054249.1 | - | 2 - 3 | 570 | 62 | |
AY059781.1 | - | 2 - 3 | 588 | 62 | ||
AY133845.1 | - | 2 - 3 | 458 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |