Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125010.3 | NP_200438.1 | 2 - 3 | 1092 | 134 | |
GenBank mRNA | AF418308.1 | - | 1 - 2 | 410 | 134 | |
AY096493.1 | - | 2 - 3 | 1004 | 134 | ||
AY123006.1 | - | 1 - 2 | 410 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |