Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125011.4 | NP_568839.1 | 8 - 9 | 945 | 130 | |
GenBank mRNA | AY048692.1 | - | 8 - 9 | 911 | 130 | |
BT025877.1 | - | 8 - 9 | 870 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |