Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037011.2 | NP_001032088.1 | 2 - 3 | 316 | 62 | |
NM_001345250.1 | NP_001329986.1 | 1 - 2 | 723 | 62 | ||
NM_001345251.1 | NP_001329984.1 | 2 - 3 | 514 | 62 | ||
NM_001345252.1 | NP_001329985.1 | 2 - 3 | 514 | 62 | ||
NM_125106.3 | NP_200534.1 | 1 - 2 | 723 | 62 | ||
GenBank mRNA | AK117264.1 | - | 1 - 2 | 274 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |