Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125258.5 | NP_200679.1 | 2 - 3 | 209 | 125 | |
GenBank mRNA | AF192490.1 | - | 2 - 3 | 167 | 125 | |
AY048227.1 | - | 2 - 3 | 158 | 125 | ||
AY086471.1 | - | 2 - 3 | 187 | 125 | ||
AY094017.1 | - | 2 - 3 | 144 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |