Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345494.1 | NP_001318860.1 | 10 - 11 | 1787 | 92 | |
NM_001345495.1 | NP_001332100.1 | 10 - 11 | 1607 | 92 | ||
NM_001345496.1 | NP_001332101.1 | 8 - 9 | 1331 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |