Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRH75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125613.3 | NP_201025.1 | 10 - 11 | 1654 | 70 | |
GenBank mRNA | AY056137.1 | - | 10 - 11 | 1580 | 70 | |
BT008581.1 | - | 10 - 11 | 1489 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |