Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125644.3 | NP_201056.1 | 6 - 7 | 872 | 61 | |
GenBank mRNA | AY084598.1 | - | 6 - 7 | 765 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |