Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103833.5 | NP_175368.2 | 16 - 17 | 1888 | 91 | |
GenBank mRNA | AF503752.1 | - | 16 - 17 | 1642 | 91 | |
AY065424.1 | - | 16 - 17 | 1786 | 91 | ||
AY094420.1 | - | 16 - 17 | 1723 | 91 | ||
BT001970.1 | - | 16 - 17 | 1642 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |