Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103909.4 | NP_564563.1 | 4 - 5 | 2151 | 69 | |
GenBank mRNA | AY091095.1 | - | 4 - 5 | 2022 | 69 | |
AY123034.1 | - | 4 - 5 | 1926 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |