Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333438.1 | NP_001322009.1 | 2 - 3 | 814 | 73 | |
NM_001333439.1 | NP_001322008.1 | 2 - 3 | 371 | 73 | ||
NM_103929.4 | NP_175463.1 | 2 - 3 | 814 | 73 | ||
GenBank mRNA | AY074314.1 | - | 2 - 3 | 738 | 73 | |
AY096416.1 | - | 2 - 3 | 427 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |