Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036783.1 | NP_001031860.1 | 2 - 3 | 670 | 128 | |
NM_001085087.2 | NP_001078556.1 | 2 - 3 | 875 | 128 | ||
NM_001125727.1 | NP_001119199.1 | 3 - 4 | 719 | 128 | ||
NM_121003.4 | NP_196528.1 | 3 - 4 | 828 | 128 | ||
GenBank mRNA | AF428373.1 | - | 3 - 4 | 621 | 128 | |
AJ131206.1 | - | 3 - 4 | 625 | 128 | ||
AY037252.1 | - | 3 - 4 | 646 | 128 | ||
AY077653.1 | - | 3 - 4 | 595 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |