Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203341.1 | NP_001190270.1 | 8 - 9 | 1040 | 66 | |
NM_001343084.1 | NP_001330616.1 | 8 - 9 | 1865 | 66 | ||
NM_001343085.1 | NP_001330617.1 | 8 - 9 | 1820 | 66 | ||
NM_001343086.1 | NP_001330618.1 | 7 - 8 | 1268 | 66 | ||
NM_121041.5 | NP_196565.1 | 8 - 9 | 1451 | 66 | ||
GenBank mRNA | AK229351.1 | - | 8 - 9 | 1194 | 66 | |
BT024896.1 | - | 7 - 8 | 939 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |