Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085094.2 | NP_001078563.1 | 3 - 4 | 416 | 137 | |
NM_001161231.2 | NP_001154703.1 | 3 - 4 | 416 | 137 | ||
NM_001203343.1 | NP_001190272.1 | 3 - 4 | 416 | 137 | ||
NM_121052.3 | NP_196576.1 | 3 - 4 | 416 | 137 | ||
GenBank mRNA | AF116527.1 | - | 3 - 4 | 416 | 137 | |
AF537203.1 | - | 3 - 4 | 307 | 137 | ||
AY850000.1 | - | 3 - 4 | 388 | 137 | ||
AY850002.1 | - | 3 - 4 | 388 | 137 | ||
AY964092.1 | - | 3 - 4 | 307 | 137 | ||
AY964093.1 | - | 3 - 4 | 307 | 137 | ||
AY964094.1 | - | 3 - 4 | 339 | 137 | ||
AY964095.1 | - | 3 - 4 | 348 | 137 | ||
AY964097.1 | - | 3 - 4 | 307 | 137 | ||
BT030637.1 | - | 3 - 4 | 307 | 137 | ||
EF547939.1 | - | 3 - 4 | 307 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |