Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR2;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121056.3 | NP_196580.1 | 7 - 8 | 1945 | 86 | |
GenBank mRNA | AB003591.1 | - | 7 - 8 | 1804 | 86 | |
AY062546.1 | - | 7 - 8 | 1818 | 86 | ||
AY093335.1 | - | 7 - 8 | 1737 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |