Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121110.2 | NP_196633.2 | 2 - 3 | 1078 | 129 | |
GenBank mRNA | DQ167579.1 | - | 2 - 3 | 1078 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |