Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343159.1 | NP_001329978.1 | 4 - 5 | 1065 | 67 | |
NM_121154.4 | NP_196677.1 | 5 - 6 | 974 | 67 | ||
GenBank mRNA | BT004028.1 | - | 5 - 6 | 724 | 67 | |
BT005077.1 | - | 5 - 6 | 500 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |