Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YUC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121170.3 | NP_196693.1 | 3 - 4 | 1273 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |