Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CESA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336002.1 | NP_001318288.1 | 2 - 3 | 427 | 121 | |
GenBank mRNA | KJ138898.1 | - | 2 - 3 | 261 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |