Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIP4;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128141.6 | NP_180152.1 | 1 - 2 | 288 | 69 | |
GenBank mRNA | AK226190.1 | - | 1 - 2 | 286 | 69 | |
BT012635.1 | - | 1 - 2 | 129 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |