Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128257.4 | NP_565632.1 | 7 - 8 | 1019 | 61 | |
GenBank mRNA | AF395058.1 | - | 6 - 7 | 816 | 61 | |
AY087622.1 | - | 7 - 8 | 904 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |