Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128508.3 | NP_180515.1 | 2 - 3 | 791 | 66 | |
GenBank mRNA | AF370568.1 | - | 2 - 3 | 666 | 66 | |
AY085710.1 | - | 2 - 3 | 772 | 66 | ||
AY090345.1 | - | 2 - 3 | 771 | 66 | ||
BT001121.1 | - | 2 - 3 | 666 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |