Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036367.2 | NP_001031444.2 | 11 - 12 | 1480 | 124 | |
NM_001202706.1 | NP_001189635.1 | 11 - 12 | 1480 | 124 | ||
NM_001336227.1 | NP_001324174.1 | 11 - 12 | 1463 | 124 | ||
NM_001336228.1 | NP_001324173.1 | 11 - 12 | 1565 | 124 | ||
NM_128543.4 | NP_029567.1 | 11 - 12 | 1499 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |