Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NADK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338499.1 | NP_001319603.1 | 9 - 10 | 1366 | 96 | |
NM_001338500.1 | NP_001327317.1 | 9 - 10 | 1262 | 96 | ||
NM_202618.3 | NP_974347.1 | 9 - 10 | 1197 | 96 | ||
GenBank mRNA | AK176132.1 | - | 9 - 10 | 1027 | 96 | |
AY383545.1 | - | 9 - 10 | 1057 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |