Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203018.1 | NP_001189947.1 | 8 - 9 | 950 | 81 | |
NM_113117.4 | NP_566700.1 | 8 - 9 | 747 | 81 | ||
GenBank mRNA | AY034923.1 | - | 8 - 9 | 738 | 81 | |
AY142571.1 | - | 8 - 9 | 610 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |